Diethyl ether stabilised, TECHNICAL, rectified
Metal drum, 195 L
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Catalog # 23807.550
Supplier: VWR Chemicals
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 4-10 ppm
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Documents
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Metal drum
- Pk:195 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



