DEET D7 (methyl D3 benzeneamide D4) 100 ng/µl 1 * 1,1 mL
Catalog # EHERXA12100010ME
Supplier: EHRENSTORFER
DEET D7 (methyl D3 benzeneamide D4) 100 ng/µl 1 * 1,1 mL
Catalog # EHERXA12100010ME
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
SafetyCaps
Supplier: S.C.A.T.
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ