ContactUsLinkComponent
FOOTREST WITH SAFELIFT HEIGHTS ADJUSTMENT FROM 100- 340 MM FOOTPLATE WITH INCLANATION ADJUSTMENT FROM 8° - 25° LANDSLIDE IMPEDING RU BBER LEVY, EPOXYDHARZBESCHICHTET, ANTHRACITE. 1 * 1 items
Catalog # BIMO9455-217
Supplier:  bimos
FOOTREST WITH SAFELIFT HEIGHTS ADJUSTMENT FROM 100- 340 MM FOOTPLATE WITH INCLANATION ADJUSTMENT FROM 8° - 25° LANDSLIDE IMPEDING RU BBER LEVY, EPOXYDHARZBESCHICHTET, ANTHRACITE. 1 * 1 items
Catalog # BIMO9455-217
Supplier:  bimos
Frequently Bought Together