ContactUsLinkComponent
Dispenser VITLAB simplex, variable, 0.5 - 5.0 mL 1 * 1 items
Catalog # VITL1601504
Supplier:  VITLAB
Dispenser VITLAB simplex, variable, 0.5 - 5.0 mL 1 * 1 items
Catalog # VITL1601504
Supplier:  VITLAB
Frequently Bought Together