ContactUsLinkComponent
Flask volumetric, capacity 25ml, wide neck, class A, amber graduated, hollow brown glass stoppered, tolerance 0.06ml, height 110mm, NS-ground 12/21, conformity certified. 1 * 2 items
Catalog # HIRS2640271
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 25ml, wide neck, class A, amber graduated, hollow brown glass stoppered, tolerance 0.06ml, height 110mm, NS-ground 12/21, conformity certified. 1 * 2 items
Catalog # HIRS2640271
Supplier:  HIRSCHMANN
Frequently Bought Together