PHOSPHODIESTERASE II TYPE I-SA FROM*BOVI NE SPLEEN 1 * 10 AU
Catalog # SIALP9041-10UN
Supplier: Merck
PHOSPHODIESTERASE II TYPE I-SA FROM*BOVI NE SPLEEN 1 * 10 AU
Catalog # SIALP9041-10UN
Supplier: Merck
Frequently Bought Together
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Glycerine, Hi-LR™
Supplier: HIMEDIA
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Ethoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals