NEURAMINIDASE TYPE X FROM*CLOSTRIDIUM PE RFRINGENS 1 * 10 AU
Catalog # SIALN2133-10UN
Supplier: Merck
NEURAMINIDASE TYPE X FROM*CLOSTRIDIUM PE RFRINGENS 1 * 10 AU
Catalog # SIALN2133-10UN
Supplier: Merck
Frequently Bought Together
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Cyclohexanone ≥99%
Supplier: Thermo Fisher Scientific