Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Cyclohexanone, Sigma-Aldrich®

25 kg
Catalog # SIALW390909-25KG
Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIALW390909-25KG
Supplier:  Merck
CAS Number:  108-94-1
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

25 kg

CAS Number:  108-94-1
Price
5 280,00 Kč
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.