3-(Methylthio)hexyl acetate, 97+% 1 * 25 g
Catalog # SIALW378917-25G-K
Supplier: Merck
Frequently Bought Together
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Iodomethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Anisole, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Xylene (mixture of isomers), extra pure
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Supplier: Honeywell Chemicals
1,5-Diaminopentane 98%
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butylboronic acid, Supelco®
Supplier: Merck