D-RIBOSE 5-PHOSPHATE DISODIUM 1 * 250 mg
Catalog # SIALR7750-250MG
Supplier: Merck
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene, EMPLURA®, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Pyridine, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Lithium perchlorate 99+% for analysis
Supplier: Thermo Fisher Scientific
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Supplier: Merck
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific