POLY(SODIUM 4-STYRENESULFONATE), MW 1,00 1 * 100 mL
Katalogové číslo SIAL527491-100ML
Supplier: Merck
POLY(SODIUM 4-STYRENESULFONATE), MW 1,00 1 * 100 mL
Katalogové číslo SIAL527491-100ML
Supplier: Merck
Frequently Bought Together
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylpentane, Sigma-Aldrich®
Supplier: Merck
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran stabilized with 300 ppm of BHT for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,3-Dihydroxynaphthalene, Supelco®
Supplier: Merck
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific