POLY(ETHYLENE GLYCOL) BEHENYL ETHER METH -ACRYLATE,50 WT% IN METHACRYLIC AC/H2O 1 * 100 mL
Catalog # SIAL468258-100ML
Supplier: Merck
POLY(ETHYLENE GLYCOL) BEHENYL ETHER METH -ACRYLATE,50 WT% IN METHACRYLIC AC/H2O 1 * 100 mL
Catalog # SIAL468258-100ML
Supplier: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ammonium peroxodisulphate (APS), Supelco®
Supplier: Merck
VWR®, Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals