ContactUsLinkComponent
MINIMUM ESSENTIAL MEDIUM EAGLE, 10×, WITH EARLE'S SALTS, WITHOUT L-GLUTAMINE AND SODIUM BICARBONATE, STERILE-FILTERED. 1 * 100 mL
Catalog # SIALM0275-100ML
Supplier:  Merck
MINIMUM ESSENTIAL MEDIUM EAGLE, 10×, WITH EARLE'S SALTS, WITHOUT L-GLUTAMINE AND SODIUM BICARBONATE, STERILE-FILTERED. 1 * 100 mL
Catalog # SIALM0275-100ML
Supplier:  Merck
Frequently Bought Together