9H-FLUORENE-9,9-DIMETHANOL, 98% 1 * 5 g
Catalog # SIAL477923-5G
Supplier: Merck
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
Toluene, GPR RECTAPUR® low in sulphur
Supplier: VWR Chemicals
n-Hexane for gas chromatography, Burdick & Jackson™
Supplier: Honeywell Chemicals
Acetone
Supplier: Lach-ner S.R.O.
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Fisher Scientific
Butyl acetate 99+%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC)
Supplier: Honeywell Chemicals
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific