ContactUsLinkComponent
MINIMUM ESSENTIAL MEDIUM EAGLE, HEPES MODIFICATION, WITH EARLE'S SALTS, L-GLUTAMINE AND 25 MM HEPES, WITHOUT SODIUM BIC 1 * 10 L
Catalog # SIALM2645-10L
Supplier:  Merck
MINIMUM ESSENTIAL MEDIUM EAGLE, HEPES MODIFICATION, WITH EARLE'S SALTS, L-GLUTAMINE AND 25 MM HEPES, WITHOUT SODIUM BIC 1 * 10 L
Catalog # SIALM2645-10L
Supplier:  Merck
Frequently Bought Together