POLYETHYLENE, ULTRA-HIGH MOLECULAR WEIGHT,SURFACE-MODIFIED, 40-48 MICRON 1 * 100 g
Catalog # SIAL434272-100G
Supplier: Merck
POLYETHYLENE, ULTRA-HIGH MOLECULAR WEIGHT,SURFACE-MODIFIED, 40-48 MICRON 1 * 100 g
Catalog # SIAL434272-100G
Supplier: Merck
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Methyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
Glycerine, Hi-LR™
Supplier: HIMEDIA
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Phenanthrene, Sigma-Aldrich®
Supplier: Merck