N-(3-(DIMETHYLAMINO)PROPYL)METHACRYLAMID E, 99% 1 * 250 mL
SIAL409472-250ML
: Merck
Frequently Bought Together
Methanol ≥99.5% NF, Ph. Eur., BP
: VWR Chemicals
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
Methyl methanesulphonate 99%
: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
Acetone, meets analytical specification of Ph.Eur.
: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
Xylene (mixture of isomers)
: Lach-ner S.R.O.
Ethanolamine, Sigma-Aldrich®
: Merck
Ethylene glycol, Sigma-Aldrich®
: Merck
Diethyl ether ≥99% stabilised for HPLC
: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals