TRIMETHYLOLPROPANE ETHOXYLATE (1 EO/OH) TRIACRYLATE, AVERAGE MN CA. 428 1 * 250 mL
Catalog # SIAL409073-250ML
Supplier: Merck
TRIMETHYLOLPROPANE ETHOXYLATE (1 EO/OH) TRIACRYLATE, AVERAGE MN CA. 428 1 * 250 mL
Catalog # SIAL409073-250ML
Supplier: Merck
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Supplier: VWR Chemicals
Aluminium foil discs
Supplier: Korff
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Klercide™ 70/30 Alcohols, Sterile
Supplier: ECOLAB CONTAMINATION CONTROL
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
4-(Dimethylamino)benzonitrile 98%
Supplier: Thermo Fisher Scientific
2-Chlorophenol ≥99%
Supplier: Thermo Fisher Scientific