INSULIN FROM FROM BOVINE PANCREAS POWDER, BIOREAGENT, SUITABLE FOR CELL CULTURE 1 * 500 mg
SIALI6634-500MG
: Merck
INSULIN FROM FROM BOVINE PANCREAS POWDER, BIOREAGENT, SUITABLE FOR CELL CULTURE 1 * 500 mg
SIALI6634-500MG
: Merck
Frequently Bought Together
Water, TECHNICAL, demineralised
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
: VWR Chemicals
Pyridine ≥99%, TECHNICAL
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals