INSULIN FROM FROM BOVINE PANCREAS POWDER, BIOREAGENT, SUITABLE FOR CELL CULTURE 1 * 250 mg
Catalog # SIALI6634-250MG
Supplier: Merck
INSULIN FROM FROM BOVINE PANCREAS POWDER, BIOREAGENT, SUITABLE FOR CELL CULTURE 1 * 250 mg
Catalog # SIALI6634-250MG
Supplier: Merck
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Pasteur pipettes, glass
Supplier: DWK Life Sciences
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals