0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
TRIMETHYLOLPROPANE TRIS(3-MERCAPTO-PROPI ONATE) 1 * 500 mL
SIAL381489-500ML
:
Merck
TRIMETHYLOLPROPANE TRIS(3-MERCAPTO-PROPI ONATE) 1 * 500 mL
SIAL381489-500ML
:
Merck
Frequently Bought Together
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
: Dr. Weigert
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Neutralising agent for the automated cleaning of glassware, neodisher® Z
: Dr. Weigert
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Dithiodiglycol (bis(2-hydroxyethyl) disulphide), Sigma-Aldrich®
: Merck
Montmorylonit K 10, powder 1kg 1 * 1 kg
: Merck