TRIMETHYLOLPROPANE TRIS(3-MERCAPTO-PROPI ONATE) 1 * 100 mL
Catalog # SIAL381489-100ML
Supplier: Merck
TRIMETHYLOLPROPANE TRIS(3-MERCAPTO-PROPI ONATE) 1 * 100 mL
Catalog # SIAL381489-100ML
Supplier: Merck
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Supplier: Dr. Weigert
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,8-Diazabicyclo[5.4.0]undec-7-ene ≥99%
Supplier: Thermo Fisher Scientific