POLY((PHENYL GLYCIDYL ETHER)-CO-FORMALDE HYDE), AVERAGE MN CA. 345 1 * 100 mL
Catalog # SIAL406775-100ML
Supplier: Merck
POLY((PHENYL GLYCIDYL ETHER)-CO-FORMALDE HYDE), AVERAGE MN CA. 345 1 * 100 mL
Catalog # SIAL406775-100ML
Supplier: Merck
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Disposable, Anti-Static Weighing Boats
Supplier: VWR Collection
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals