2-DIMETHYLAMINO-2-METHYL-1-PROPANOL, TEC H., 80% SOLUTION IN WATER 1 * 100 mL
Catalog # SIAL423041-100ML
Supplier: Merck
2-DIMETHYLAMINO-2-METHYL-1-PROPANOL, TEC H., 80% SOLUTION IN WATER 1 * 100 mL
Catalog # SIAL423041-100ML
Supplier: Merck
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Supplier: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
n-Heptane 95%
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific