DIETHYLPHOSPHORAMIDOUS DICHLORIDE, 97% 1 * 5 g
Catalog # SIAL362891-5G
Supplier: Merck
Frequently Bought Together
Xylen (směs izomerů) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyklohexan, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
n-Heptan ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
terc-Butylmethylether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitril s 0,1% (v/v) Kyselina mravenčí, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Klercide™ 70/30 Alcohols, Sterile
Supplier: ECOLAB CONTAMINATION CONTROL
N,N-Dimethylformamid, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Sodium triacetoxyborohydride 95%
Supplier: Apollo Scientific
Dichlormethan ≥99.8% (by GC) stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals