LITHIUM DIISOPROPYLAMIDE, 2.0M SOLUTION IN HEPTANE/THF/ETHYLBENZENE 1 * 100 mL
Catalog # SIAL361798-100ML
Supplier: Merck
LITHIUM DIISOPROPYLAMIDE, 2.0M SOLUTION IN HEPTANE/THF/ETHYLBENZENE 1 * 100 mL
Catalog # SIAL361798-100ML
Supplier: Merck
Frequently Bought Together
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
tert-Butanol ACS
Supplier: Thermo Fisher Scientific
1,3-Propanediol ≥99%
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals