3-ISOPROPENYL-ALPHA,ALPHA-DIMETHYLBENZYL ISOCYANATE, 95% 1 * 250 mL
Catalog # SIAL361771-250ML
Supplier: Merck
3-ISOPROPENYL-ALPHA,ALPHA-DIMETHYLBENZYL ISOCYANATE, 95% 1 * 250 mL
Catalog # SIAL361771-250ML
Supplier: Merck
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Butyl acrylate stabilised, Sigma-Aldrich®
Supplier: Merck
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
ISODECYL METHACRYLATE, 97.5+% 1 * 1 L
Supplier: Merck