4,4'-METHYLENEBIS(2-METHYLCYCLOHEXYL-AMI NE), 99%, MIXTURE OF ISOMERS 1 * 250 mL
SIAL369500-250ML
: Merck
4,4'-METHYLENEBIS(2-METHYLCYCLOHEXYL-AMI NE), 99%, MIXTURE OF ISOMERS 1 * 250 mL
SIAL369500-250ML
: Merck
Frequently Bought Together
o-Xylene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Trifluoroacetic acid ≥99.9% for peptide synthesis
: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
: VWR Chemicals
Glycerine ≥97%, TECHNICAL
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Cyclohexane for pesticide residue analysis
: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific