Ethynylmagnesium bromide solution, 0.5M tetrahydrofuran 1 * 800 mL
Catalog # SIAL346152-800ML
Supplier: Merck
Ethynylmagnesium bromide solution, 0.5M tetrahydrofuran 1 * 800 mL
Catalog # SIAL346152-800ML
Supplier: Merck
Frequently Bought Together
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) trifluoroacetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific