ANTI-MOUSE IGG (FAB SPECIFIC) FITC*CONJU GATE ANTIBO 1 * 0,5 mL
Catalog # SIALF5262-.5ML
Supplier: Merck
ANTI-MOUSE IGG (FAB SPECIFIC) FITC*CONJU GATE ANTIBO 1 * 0,5 mL
Catalog # SIALF5262-.5ML
Supplier: Merck
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Goat serum
Supplier: Biowest
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF