SODIUM CYANOBOROHYDRIDE, 1.0M SOLUTION I N TETRAHYDROFURAN 1 * 100 mL
Catalog # SIAL296813-100ML
Supplier: Merck
SODIUM CYANOBOROHYDRIDE, 1.0M SOLUTION I N TETRAHYDROFURAN 1 * 100 mL
Catalog # SIAL296813-100ML
Supplier: Merck
Frequently Bought Together
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diiodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexanone ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Bromosuccinimide 99%
Supplier: Thermo Fisher Scientific
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Cesium fluoride 99.9%, extra pure
Supplier: Thermo Fisher Scientific
Formaldehyde 37% (w/w) in aqueous solution stabilised
Supplier: Thermo Fisher Scientific