CIS-11-EICOSENOIC ACID METHYL ESTER 1 * 25 mg
Catalog # SIALE6885-25MG
Dodavatel: Merck
Frequently Bought Together
Glycerine ≥98%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Diethyl carbonate for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Xylene (mixture of isomers) 99% ACS
Dodavatel: Thermo Fisher Scientific
4-Methyl-2-pentanone 99+% for HPLC
Dodavatel: Thermo Fisher Scientific
Methanol 99+%, extra pure
Dodavatel: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for HPLC
Dodavatel: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Dodavatel: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Methanol, J.T.Baker®
Dodavatel: Avantor
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Dodavatel: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Dodavatel: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals