CIS-11-EICOSENOIC ACID METHYL ESTER 1 * 100 mg
Katalogové číslo SIALE6885-100MG
Dodavatel: Merck
Frequently Bought Together
n-Nonane 99%, Reagent Grade
Dodavatel: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Dodavatel: Thermo Fisher Scientific
2-Butoxyethanol 99%, extra pure
Dodavatel: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Dodavatel: Thermo Fisher Scientific
Cyclohexane ≥99%
Dodavatel: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Dodavatel: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
Dodavatel: Honeywell Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Dodavatel: Thermo Fisher Scientific
Acetone 99.8% for HPLC
Dodavatel: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals