4-(METHYLAMINO)BUTYRIC ACID HYDROCHLORID E, 99% 1 * 100 g
Katalogové číslo SIAL263710-100G
Supplier: Merck
4-(METHYLAMINO)BUTYRIC ACID HYDROCHLORID E, 99% 1 * 100 g
Katalogové číslo SIAL263710-100G
Supplier: Merck
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.002% H₂O) ≥99.8%
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
n-Heptane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ferrozine mono-sodium salt hydrate, Supelco®
Supplier: Merck
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific