4-NITROPHENOL SOLUTION, 10MM 1 * 100 mL
Catalog # SIALN7660-100ML
Supplier: Merck
Frequently Bought Together
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Supplier: Dr. Weigert
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Sulphur trioxide pyridine complex for synthesis
Supplier: Merck
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
dsDNA quantitation kits, AccuBlue® and AccuClear®
Supplier: Biotium
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
Culture bottles and culture flasks, DURAN®
Supplier: DWK Life Sciences
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals