VINYLMAGNESIUM BROMIDE, 1.0M SOLUTION IN TETRAHYDROFURAN 1 * 100 mL
Catalog # SIAL225584-100ML
Dodavatel: Merck
VINYLMAGNESIUM BROMIDE, 1.0M SOLUTION IN TETRAHYDROFURAN 1 * 100 mL
Catalog # SIAL225584-100ML
Dodavatel: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Dodavatel: Merck
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Dodavatel: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals