TRIS(HYDROXYMETHYL)AMINOMETHANE, 99.8+%, 1 * 100 g
Catalog # SIAL252859-100G
Supplier: Merck
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Pentan 99+%, extra pure
Supplier: Thermo Fisher Scientific
[EN]METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Acetonitril 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Benzonitril 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
[EN]TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
2-methylbutan 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridin ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilizovaný for spectroscopy
Supplier: Thermo Fisher Scientific
izo-Amylalkohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Supplier: PanReac AppliChem