DOWEX 1X8-200 ION-EXCHANGE RESIN 1 * 2,5 kg
Catalog # SIAL217425-2.5KG
Supplier: Merck
Frequently Bought Together
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
n-Hexane, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
1-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9%, extra pure low in benzene
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
HCL ACID REAGENT 1 * 20 L
Supplier: Hach