Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MOUSE MONOCLONAL ANTI-HUMAN IGG1CLONE 8C /6-39 BIOT 1 * 0,5 mL
Catalog # SIALB6775-.5ML
Supplier:
Merck
Expand image
MOUSE MONOCLONAL ANTI-HUMAN IGG1CLONE 8C /6-39 BIOT 1 * 0,5 mL
Catalog # SIALB6775-.5ML
Supplier:
Merck
Frequently Bought Together
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection