POTASSIUM HYDRIDE, 30 WT. % DISPERSION I N MINERAL OIL 1 * 75 g
Catalog # SIAL215813-75G
Supplier: Merck
POTASSIUM HYDRIDE, 30 WT. % DISPERSION I N MINERAL OIL 1 * 75 g
Catalog # SIAL215813-75G
Supplier: Merck
Frequently Bought Together
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Copper(II) sulphate pentahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Magnesium nitrate hexahydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier: VWR Chemicals