(+)-DIHYDROCARVONE, 98%, MIXTURE OF ISOM ERS 1 * 25 g
Catalog # SIAL218286-25G
Supplier: Merck
Frequently Bought Together
Acetone reagent ≥99.5% ACS
Supplier: Merck
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers)
Supplier: Lach-ner S.R.O.
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Diethyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific