Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TRIS(2-METHOXYETHOXY)VINYLSILANE, 98% 1 * 100 mL
Catalog # SIAL175587-100ML
Supplier:
Merck
Expand image
TRIS(2-METHOXYETHOXY)VINYLSILANE, 98% 1 * 100 mL
Catalog # SIAL175587-100ML
Supplier:
Merck
Frequently Bought Together
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N<sub>α</sub>-(2,4-Dinitro-5-fluorophenyl)-L-valinamide ≥98.0%, LiChropur™ for chiral derivatisation, Supelco®
Supplier: Merck
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific