POLY(1,4-BUTYLENE ADIPATE), AVERAGE MW C A. 12,000 (GPC) 1 * 25 g
Catalog # SIAL181501-25G
Supplier: Merck
POLY(1,4-BUTYLENE ADIPATE), AVERAGE MW C A. 12,000 (GPC) 1 * 25 g
Catalog # SIAL181501-25G
Supplier: Merck
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
P-TOLUIC ACID, 98% 1 * 5 g
Supplier: Merck
Trichloroethylene 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Succinonitrile ≥98%
Supplier: Thermo Fisher Scientific
Xanthone ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Iodomethane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific