Barium Standard for ICP 1 * 100 mL
Catalog # SUPL59943-100ML
Supplier: Merck
Frequently Bought Together
Butane-1,3-diol, Reference Standard
Supplier: USP
pH buffers, ready to use
Supplier: WTW
Single element standards for ICP-OES, copper
Supplier: Agilent
Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Pesticide reference standard, Dieldrin 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Ammonium molybdate solution R4 Reag. Ph. Eur. 1005705
Supplier: VWR Chemicals
Silicium Standard for ICP, TraceCERT®, Supelco®
Supplier: Merck
Buffer Solutions
Supplier: Thermo Fisher Scientific
Supelco® Organic Reference Standard, Luteolin 7-glucoside
Supplier: Merck
HPTLC CALIBRATION MIX 1 * 1 mL
Supplier: Merck
(±)-Camphene
Supplier: Merck
Accessories for 410
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Dessiccant, Reference Standards
Supplier: USP
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals