Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 6×1 L, 6 L
  33665-6X1L
Buffer solutions, colour coded, Fluka™
  33665-6X1L
 :  
Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, Plastic bottle, 6×1 L, 6 L

 :  
 
12 600,00 Kč
   
1 * 6 L
 

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

 

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • Colour:
    Red
  • Pk Info:
    6×1 L
  • pH value:
    4,00