TRIS(HYDROXYMETHYL)NITROMETHANE, 98% 1 * 100 g
Catalog # SIAL108189-100G
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
o-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals