Orders LinkContactUsLinkComponent
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
14/23 NS, 10/19 NS
Catalog # 201-0639
Supplier:   SciLabware
Adapters, reduction, Quickfit®
Catalog # 201-0639
Supplier:  SciLabware
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
14/23 NS, 10/19 NS
Price
1 590,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    10/19 NS