Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
HPLC columns, LiChrospher®

HPLC columns, LiChrospher®

Analytical, RP-select B, 60 Å, 5 µm, 4,60 mm, 100 mm
Catalog # 1.50573.0001
Supplier:   Merck
HPLC columns, LiChrospher®
Catalog # 1.50573.0001
Supplier:  Merck
HPLC columns, LiChrospher®
HPLC columns, LiChrospher®

Analytical, RP-select B, 60 Å, 5 µm, 4,60 mm, 100 mm

Price
19 960,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents

LiChrospher® is Merck's reliable and versatile, traditionally-produced, spherical silica. LiChrospher® is available with different modifications.

LiChrospher® Si 60 and Si 100 are versatile HPLC spherical silica sorbents for normal phase chromatography.
LiChrospher® CN has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. In addition, selective charged interactions makes it even more versatile.
LiChrospher® NH₂ possesses polar and hydrophobic properties and can be used for normal phase, reversed phase and ion exchange chromatography. Typical applications are the separations of carbohydrates by reversed phase chromatography or the separation of nucleotides by ion exhange chromatography.

LiChrospher® DIOL has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. LiChrospher® DIOL is also suitable for exclusion chromatography.
LiChrospher® RP-select B is a highly reproducible, traditionally-produced spherical silica gel with reversed phase properties. This sorbent is ideal for the separation of basic compounds, but can also be used with neutral and acidic samples.

LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally-produced spherical silica gel sorbents with reversed-phase properties. They are ideal for the separation of neutral, acidic and weak basic compounds.
LiChrospher® WP 300 RP-18 is a highly selective and reliable HPLC column for the separation of peptides and low molecular weight proteins. LiChrospher® WP 300 RP-18 enables the tailing-free separation of basic compounds, and is suitable for the separation of tRNA.
LiChrospher® PAH is a highly efficient and selective HPLC column, especially designed for the high resolution separation of 16 PAH (polycyclic aromatic hydrocarbons) according to EPA 610 and 550, plus benzo(e)pyrene and perylene.

LiChroCART® cartridges (2, 3, 4, 4,6 mm I-Ø) require the use of manu-CART® cartridge holder 1.51486.0001
LiChroCART® cartridges (10×250 mm) require the use of manu-CART® cartridge holder 1.51419.0001
Guard cartridges have to be connected to the LiChroCART® cartridges using manu-CART® cartridge holder 1.51486.0001.

LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally produced spherical silica gel sorbents with reversed phase properties. They are well suited for the separation of neutral, acidic and weak basic compounds.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    60 Å
  • Length:
    100 mm
  • Seq:
    78
  • Phase:
    RP-select B
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.