Cyclohexanone, Sigma-Aldrich®
Glass bottle, 2,5 L
Catalog # SIALC102180-2.5L
Supplier: Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIALC102180-2.5L
Supplier: Merck
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C
- UN:1915
- ADR:3,III
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:203-631-1
- REACH:01-2119453616-35
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:SIALC102180-2.5L
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



