Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Cyclohexanone, Sigma-Aldrich®

Cyclohexanone, Sigma-Aldrich®

Glass bottle, 500 mL
Catalog # SIAL398241-500ML
Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Sigma-Aldrich®
Catalog # SIAL398241-500ML
Supplier:  Merck
CAS Number:  108-94-1
Cyclohexanone, Sigma-Aldrich®
Cyclohexanone, Sigma-Aldrich®

Glass bottle, 500 mL

CAS Number:  108-94-1
Price
1 480,00 Kč
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications